1-Benzyl-3-piperidone hydrochloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

B3419-25G B3419-5G
TCB3419-25GEA 164.32 CAD
TCB3419-25G TCB3419-5G
1-Benzyl-3-piperidone hydrochloride ≥97.0% (by HPLC, titration analysis)
1-Benzyl-3-piperidone hydrochloride

CAS Number: 50606-58-1 MDL Number: MFCD00150096 Molecular Formula: C12H15NO Molecular Weight: 225.72 Purity/Analysis Method: 97.0% (HPLC,T) Form: Crystal Boiling point (°C): 91 Melting point (°C): 171

Formula: C₁₂H₁₅NO·HCl
Boiling Pt: 91 °C
Melting Pt: 171 °C
MDL Number: MFCD00012791
CAS Number: 50606-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cab99f6fb16a8bba65bac47848584d6cac607d74&vid=cab99f6fb16a8bba65bac47848584d6cac607d74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR