2-Bromo-5-nitroanisole ≥98.0%

Supplier: TCI America

B3418-25G B3418-5G
TCB3418-25GEA 485.06 CAD
TCB3418-25G TCB3418-5G
2-Bromo-5-nitroanisole ≥98.0%
2-Bromo-5-nitroanisole

CAS Number: 77337-82-7
MDL Number: MFCD00041250
Molecular Formula: C7H6BrNO3
Molecular Weight: 232.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 104
Lambda max.: 324 nm (EtOH)


Other Product Names / Synonyms: 1-Bromo-2-methoxy-4-nitrobenzene

Formula: C₇H₆BrNO₃
Melting Pt: 104 °C
MDL Number: MFCD00041250
CAS Number: 77337-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c23fc1f033550751a8d7a64bb4e70d905db2dd0&vid=bcccfd68a72297d8493911dba1528e8c25207294&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR