1,4-Benzodioxane-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-2-carboxylic acid

B3406-25G B3406-5G
TCB3406-25GEA 216.46 CAD
TCB3406-25G TCB3406-5G
1,4-Benzodioxane-2-carboxylic acid ≥98.0% (by GC, titration analysis)
1,4-Benzodioxane-2-carboxylic acid

CAS Number: 3663-80-7
MDL Number: MFCD00084888
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 126
Lambda max.: 282 nm (MeOH)

Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00084888
CAS Number: 3663-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8aa515f3852b2546cc8cd6f871c4bec1b90cf7ee&vid=8aa515f3852b2546cc8cd6f871c4bec1b90cf7ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR