1-Benzylimidazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B3387-25G
B3387-5G
TCB3387-25GEA
102.7
CAD
TCB3387-25G
TCB3387-5G
1-Benzylimidazole ≥98.0% (by titrimetric analysis)
1-Benzylimidazole
CAS Number: 4238-71-5
MDL Number: MFCD00005296
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 122
Melting point (°C): 73
Lambda max.: 215 nm (EtOH)
Formula:
C₁₀H₁₀N₂ Boiling Pt: 122 °C Melting Pt: 73 °C |
MDL Number:
MFCD00005296 CAS Number: 4238-71-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...