4'-Bromo-3'-nitroacetophenone ≥98.0%
Supplier: TCI America
Synonyms:
1-(4-Bromo-3-nitrophenyl)ethan-1-one, 1-(4-Bromo-3-nitrophenyl)ethanone
B3383-1G
B3383-5G
TCB3383-1GEA
44.24
CAD
TCB3383-1G
TCB3383-5G
4'-Bromo-3'-nitroacetophenone ≥98.0%
4'-Bromo-3'-nitroacetophenone
CAS Number: 18640-58-9
MDL Number: MFCD00016985
Molecular Formula: C8H6BrNO3
Molecular Weight: 244.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 119
Formula:
C₈H₆BrNO₃ Melting Pt: 119 °C |
MDL Number:
MFCD00016985 CAS Number: 18640-58-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...