(1H,3H)-Quinazoline-2,4-dione ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3381-5G B3381-25G
TCB3381-5GEA 101.12 CAD
TCB3381-5G TCB3381-25G
(1H,3H)-Quinazoline-2,4-dione ≥98.0% (by HPLC, titration analysis)
(1H,3H)-Quinazoline-2,4-dione

CAS Number: 86-96-4
MDL Number: MFCD00006699
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 354
Lambda max.: 217 nm (EtOH)


Other Product Names / Synonyms: 2,4(1H,3H)-Quinazolinedione

Formula: C₈H₆N₂O₂
Melting Pt: 354 °C
MDL Number: MFCD00006699
CAS Number: 86-96-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR