1-Boc-piperidine-3-carboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B3376-25G B3376-5G
TCB3376-25GEA 248.06 CAD
TCB3376-25G TCB3376-5G
1-Boc-piperidine-3-carboxylic acid ≥97.0% (by GC, titration analysis)
1-Boc-piperidine-3-carboxylic acid

CAS Number: 84358-12-3
MDL Number: MFCD00673775
Molecular Formula: C11H19NO4
Molecular Weight: 229.28
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 160


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Boc-nipecotic Acid, 1-(tert-Butoxycarbonyl)nipecotic Acid, 1-Boc-3-piperidinecarboxylic Acid

Formula: C₁₁H₁₉NO₄
Melting Pt: 160 °C
MDL Number: MFCD00673775
CAS Number: 84358-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3f0d39ab22ed8922935d8ee91823e63119ae8ba&vid=b3f0d39ab22ed8922935d8ee91823e63119ae8ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR