1,2-Bis(diphenylphosphino)benzene ≥98.0%

Supplier: TCI America

B3372-1G B3372-5G
TCB3372-1GEA 140.62 CAD
TCB3372-1G TCB3372-5G
1,2-Bis(diphenylphosphino)benzene ≥98.0%
1,2-Bis(diphenylphosphino)benzene

CAS Number: 13991-08-7
MDL Number: MFCD00014081
Molecular Formula: C30H24P2
Molecular Weight: 446.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 188


Product Categories: Synthetic Organic Chemistry, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: dppbz

Formula: C₃₀H₂₄P₂
Melting Pt: 188 °C
MDL Number: MFCD00014081
CAS Number: 13991-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5290b74c660a6379e5d19eefafd47c419148e9c5&vid=5290b74c660a6379e5d19eefafd47c419148e9c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR