3-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3370-1G B3370-5G
TCB3370-1GEA 60.04 CAD
TCB3370-1G TCB3370-5G
3-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-5-nitrobenzoic acid

CAS Number: 6307-83-1
MDL Number: MFCD00100098
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 162

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00100098
CAS Number: 6307-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19dba17a4f8a60d82e5811bd3e124cf466740f24&vid=19dba17a4f8a60d82e5811bd3e124cf466740f24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR