2-Bromo-5-chlorobenzotrifluoride ≥98.0%

Supplier: TCI America

B3366-25G B3366-5G
TCB3366-25GEA 140.46 CAD
TCB3366-25G TCB3366-5G
2-Bromo-5-chlorobenzotrifluoride ≥98.0%
2-Bromo-5-chlorobenzotrifluoride

CAS Number: 344-65-0
MDL Number: MFCD00010308
Molecular Formula: C7H3BrClF3
Molecular Weight: 259.45
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Melting point (°C): 19
Flash Point (°C): 99
Specific Gravity (20/20): 1.77


Other Product Names / Synonyms: 1-Bromo-4-chloro-2-(trifluoromethyl)benzene, 2-Bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₃BrClF₃
Boiling Pt: 198 °C
Melting Pt: 19 °C
Density: 1.77 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00010308
CAS Number: 344-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b3ceefc7000a3dfe2ac602338c2343ae780533b&vid=0b3ceefc7000a3dfe2ac602338c2343ae780533b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR