4'-Benzyloxyacetophenone ≥98.0%
Supplier: TCI America
B3365-1G
B3365-5G
TCB3365-1GEA
26.86
CAD
TCB3365-1G
TCB3365-5G
4'-Benzyloxyacetophenone ≥98.0%
4'-Benzyloxyacetophenone
CAS Number: 54696-05-8
MDL Number: MFCD00017247
Molecular Formula: C15H14O2
Molecular Weight: 226.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 93
Formula:
C₁₅H₁₄O₂ Melting Pt: 93 °C |
MDL Number:
MFCD00017247 CAS Number: 54696-05-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...