3-Bromo-5-methylpyridin-2-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Bromo-2-hydroxy-5-methylpyridine

B3350-1G B3350-5G
TCB3350-1GEA 287.56 CAD
TCB3350-1G TCB3350-5G
3-Bromo-5-methylpyridin-2-ol ≥98.0% (by GC, titration analysis)
3-Bromo-5-methylpyridin-2-ol

CAS Number: 17282-02-9
MDL Number: MFCD03791312
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 161


Other Product Names / Synonyms: 3-Bromo-5-methyl-2-pyridinol, 3-Bromo-2-hydroxy-5-methylpyridine

Formula: C6H6BrNO
MW: 188.02 g/mol
Melting Pt: 161 °C
MDL Number: MFCD03791312
CAS Number: 17282-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1bad32aa1e9818182135e61c5b4b00539cb653a&vid=f1bad32aa1e9818182135e61c5b4b00539cb653a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR