1-Bromo-2-chloro-4-fluorobenzene ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-3-chloro-1-fluorobenzene, 2-Chloro-4-fluorobromobenzene, 4-Bromo-3-chlorofluorobenzene

B3349-25G B3349-5G
TCB3349-25GEA 169.06 CAD
TCB3349-25G TCB3349-5G
1-Bromo-2-chloro-4-fluorobenzene ≥98.0%
1-Bromo-2-chloro-4-fluorobenzene

CAS Number: 110407-59-5
MDL Number: MFCD00142587
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.75

Formula: C₆H₃BrClF
MW: 209.44 g/mol
Density: 1.75 (20 °C)
MDL Number: MFCD00142587
CAS Number: 110407-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d45af4e735666560211f0e735a8d6c1f8de6356&vid=6d45af4e735666560211f0e735a8d6c1f8de6356&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR