4-Bromomethyl-1,2-dihydroquinoline-2-one ≥97.0%

Supplier: TCI America

Synonyms: 4-(Bromomethyl)quinolin-2(1H)-one

B3345-25G B3345-5G
TCB3345-25GEA 148.84 CAD
TCB3345-25G TCB3345-5G
4-Bromomethyl-1,2-dihydroquinoline-2-one ≥97.0%
4-Bromomethyl-1,2-dihydroquinoline-2-one

CAS Number: 4876-10-2
MDL Number: MFCD03426152
Molecular Formula: C10H8BrNO
Molecular Weight: 238.08
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 257


Other Product Names / Synonyms: 4-(Bromomethyl)-2-quinolinol

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
Melting Pt: 257 °C
MDL Number: MFCD03426152
CAS Number: 4876-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98e6f7ba37582c85a193705f895633be7419f521&vid=98e6f7ba37582c85a193705f895633be7419f521&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR