4-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromo-2-methylbenzoic acid

B3337-25G B3337-5G
TCB3337-25GEA 1017.52 CAD
TCB3337-25G TCB3337-5G
4-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)
4-Bromo-o-toluic acid

CAS Number: 68837-59-2
MDL Number: MFCD00040905
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 181


Other Product Names / Synonyms: 4-Bromo-o-toluic Acid

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00040905
CAS Number: 68837-59-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1f77f69c44b99ab94c509448aea72054cbf909a&vid=290d655284aa25b0046778d01238d32295ac9a83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR