3-Bromo-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3336-25G B3336-5G
TCB3336-25GEA 124.82 CAD
TCB3336-25G TCB3336-5G
3-Bromo-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-4-methoxybenzoic acid

CAS Number: 99-58-1
MDL Number: MFCD00020295
Molecular Formula: C8H7BrO3
Molecular Weight: 231.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 222


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 3-Bromo-p-anisic Acid

Formula: C₈H₇BrO₃
Melting Pt: 222 °C
MDL Number: MFCD00020295
CAS Number: 99-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebd3a1a15b62ca2222463b08771caafddce56742&vid=25c3bc47fe46843914bf8ac24d3a314f1ecd827e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR