4-(Boc-amino)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3331-5G
B3331-25G
TCB3331-5GEA
139.04
CAD
TCB3331-5G
TCB3331-25G
4-(Boc-amino)pyridine ≥98.0% (by GC, titration analysis)
4-(Boc-amino)pyridine
CAS Number: 98400-69-2
MDL Number: MFCD02179232
Molecular Formula: C10H14N2O2
Molecular Weight: 194.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 150
Other Product Names / Synonyms: 4-(Boc-amino)pyridine, tert-Butyl N-(4-Pyridyl)carbamate
Formula:
C₁₀H₁₄N₂O₂ Melting Pt: 150 °C |
MDL Number:
MFCD02179232 CAS Number: 98400-69-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...