Boc-Aib-OH ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3323-25G B3323-5G
TCB3323-25GEA 121.66 CAD
TCB3323-25G TCB3323-5G
Boc-Aib-OH ≥98.0% (by GC, titration analysis)
Boc-Aib-OH

CAS Number: 30992-29-1
MDL Number: MFCD00042973
Molecular Formula: C9H17NO4
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 120


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 2-(Boc-amino)isobutyric Acid, N-Boc-alpha-methylalanine, Boc-Aib-OH

Formula: C₉H₁₇NO₄
Melting Pt: 120 °C
MDL Number: MFCD00042973
CAS Number: 30992-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b60ad7ffbbc2e4d2935f1a48bac7e03976bba360&vid=b60ad7ffbbc2e4d2935f1a48bac7e03976bba360&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR