3-Bromo-4-chlorobenzoic acid ≥98.0%

Supplier: TCI America

B3308-1G B3308-5G
TCB3308-1GEA 54.51 CAD
TCB3308-1G TCB3308-5G
3-Bromo-4-chlorobenzoic acid ≥98.0%
3-Bromo-4-chlorobenzoic acid

CAS Number: 42860-10-6
MDL Number: MFCD00079706
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 220

Formula: C₇H₄BrClO₂
Melting Pt: 220 °C
MDL Number: MFCD00079706
CAS Number: 42860-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64593b1ff4f4daa1db9dc19d8153400d718c503e&vid=64593b1ff4f4daa1db9dc19d8153400d718c503e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR