2-Bromo-5-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid

B3307-1G B3307-5G
TCB3307-1GEA 120.71 CAD
TCB3307-1G TCB3307-5G
2-Bromo-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Bromo-5-pyridineboronic acid

CAS Number: 223463-14-7
MDL Number: MFCD03411558
Molecular Formula: C5H5BBrNO2
Molecular Weight: 201.81
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 165


Other Product Names / Synonyms: 6-Bromo-3-pyridylboronic Acid

Formula: C₅H₅BBrNO₂
MW: 201.81
Melting Pt: 165 °C
MDL Number: MFCD03411558
CAS Number: 223463-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc2c01d1d4cbedc0e6e83a4dcc198e8f9ccb870a&vid=cc2c01d1d4cbedc0e6e83a4dcc198e8f9ccb870a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR