4-Bromo-3,5-dimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3306-5G B3306-25G
TCB3306-5GEA 39.5 CAD
TCB3306-5G TCB3306-25G
4-Bromo-3,5-dimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Bromo-3,5-dimethyl-1H-pyrazole

CAS Number: 3398-16-1
MDL Number: MFCD00005242
Molecular Formula: C5H7BrN2
Molecular Weight: 175.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123

Formula: C₅H₇BrN₂
Melting Pt: 123 °C
MDL Number: MFCD00005242
CAS Number: 3398-16-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR