4-Bromo-2-methylpyridine ≥98.0%

Supplier: TCI America

B3279-1G B3279-5G
TCB3279-1GEA 58.46 CAD
TCB3279-1G TCB3279-5G
4-Bromo-2-methylpyridine ≥98.0%
4-Bromo-2-methylpyridine

CAS Number: 22282-99-1
MDL Number: MFCD03788196
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 79
Specific Gravity (20/20): 1.50


Other Product Names / Synonyms: 4-Bromo-2-picoline

Formula: C₆H₆BrN
MW: 172.02
Boiling Pt: 76 °C
Density: 1.50 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD03788196
CAS Number: 22282-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22cb8f3b16af1eea80963091534323f4b7b2038b&vid=22cb8f3b16af1eea80963091534323f4b7b2038b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR