N-(tert-Butoxycarbonyl)-L-2-phenylglycinol ≥98.0%
Supplier: TCI America
Synonyms:
Boc-Phenylglycinol, (S)-tert-Butyl (2-hydroxy-1-phenylethyl)carbamate, N-Boc-L-2-phenylglycinol, (S)-2-(tert-Butoxycarbonylamino)-2-phenylethanol
CAS Number: 117049-14-6
MDL Number: MFCD00274206
Molecular Formula: C13H19NO3
Molecular Weight: 237.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 136
Specific rotation [a]20/D: 35.5 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-L-2-phenylglycinol, (S)-2-(tert-Butoxycarbonylamino)-2-phenylethanol
Formula:
C₁₃H₁₉NO₃ MW: 237.30 g/mol Melting Pt: 136 °C |
MDL Number:
MFCD00274206 CAS Number: 117049-14-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...