(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol ≥98.0%

Supplier: TCI America

Synonyms: Boc-D-Phenylglycinol, (-)-N-Boc-D-α-phenylglycinol

B3270-1G B3270-5G
TCB3270-1GEA 39.5 CAD
TCB3270-1G TCB3270-5G
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol ≥98.0%
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol

CAS Number: 102089-74-7
MDL Number: MFCD00274205
Molecular Formula: C13H19NO3
Molecular Weight: 237.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 137
Specific rotation [a]20/D: -38 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-D-2-phenylglycinol, (R)-2-(tert-Butoxycarbonylamino)-2-phenylethanol

Formula: C₁₃H₁₉NO₃
MW: 237.3 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00274205
CAS Number: 102089-74-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db44b73d49a9740349e96a17661bb0d823112e31&vid=db44b73d49a9740349e96a17661bb0d823112e31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR