2-Bromo-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3243-25G
B3243-5G
TCB3243-25GEA
301.78
CAD
TCB3243-25G
TCB3243-5G
2-Bromo-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
2-Bromo-N,N-dimethylaniline
CAS Number: 698-00-0
MDL Number: MFCD00013522
Molecular Formula: C8H10BrN
Molecular Weight: 200.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 108
Specific Gravity (20/20): 1.39
Formula:
C₈H₁₀BrN Boiling Pt: 108 °C Density: 1.39 (20 °C) |
MDL Number:
MFCD00013522 CAS Number: 698-00-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...