1-Boc-piperidine-4-carboxylic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America

B3241-5G B3241-25G
TCB3241-5GEA 83.74 CAD
TCB3241-5G TCB3241-25G
1-Boc-piperidine-4-carboxylic acid ≥96.0% (by GC, titration analysis)
1-Boc-piperidine-4-carboxylic acid

CAS Number: 84358-13-4
MDL Number: MFCD00076999
Molecular Formula: C11H19NO4
Molecular Weight: 229.28
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 152


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Boc-isonipecotic Acid, 1-(tert-Butoxycarbonyl)isonipecotic Acid, 1-Boc-4-piperidinecarboxylic Acid

Formula: C₁₁H₁₉NO₄
Melting Pt: 152 °C
MDL Number: MFCD00076999
CAS Number: 84358-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7effe94b45b21cd7e51f524d3a4810580097f2e4&vid=7effe94b45b21cd7e51f524d3a4810580097f2e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR