N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98,0

Supplier: TCI America

Synonyms: N-Boc-D-2-phenylglycine, BOC-D-alpha-phenylglycine, Boc-D-Phg-OH

B3230-1G B3230-5G
TCB3230-1GEA 28.44 CAD
TCB3230-1G TCB3230-5G
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98,0
N-(tert-Butoxycarbonyl)-D-2-phenylglycine

CAS Number: 33125-05-2
MDL Number: MFCD00062043
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 91
Specific rotation [a]20/D: -140 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-2-phenylglycine, Boc-D-Phg-OH

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00062043
CAS Number: 33125-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81e9319d47228339ac75b75212642bc51f177ef9&vid=81e9319d47228339ac75b75212642bc51f177ef9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR