Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥96.0%

Supplier: TCI America

Synonyms: Pd(DIPHOS)2, Pd(dppe)2

B3224-1G B3224-5G
TCB3224-1GEA 150.95 CAD
TCB3224-1G TCB3224-5G
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥96.0%
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)

CAS Number: 31277-98-2
MDL Number: MFCD00009880
Molecular Formula: C52H48P4Pd
Molecular Weight: 903.27
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Storage Temperature: >-20°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Pd(DIPHOS)2, Pd(dppe)2

Formula: C₅₂H₄₈P₄Pd
MW: 903.27 g/mol
MDL Number: MFCD00009880
CAS Number: 31277-98-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b66fb120711b84256bf3d0f2d324e65ce41e6250&vid=b66fb120711b84256bf3d0f2d324e65ce41e6250&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR