4-Bromo-2,6-difluoroaniline ≥98.0%

Supplier: TCI America

B3220-1G B3220-5G B3220-25G
TCB3220-1GEA 104.28 CAD
TCB3220-1G TCB3220-5G TCB3220-25G
4-Bromo-2,6-difluoroaniline ≥98.0%
4-Bromo-2,6-difluoroaniline

CAS Number: 67567-26-4
MDL Number: MFCD00013389
Molecular Formula: C6H4BrF2N
Molecular Weight: 208.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 64

Formula: C₆H₄BrF₂N
MW: 208.01
Melting Pt: 64 °C
MDL Number: MFCD00013389
CAS Number: 67567-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebf90c030763ec63bae3175aee15e71df9960dfd&vid=ebf90c030763ec63bae3175aee15e71df9960dfd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR