N-(tert-Butoxycarbonyl)-L-2-phenylglycine 98,0

Supplier: TCI America

Synonyms: Boc-Phg-OH, BOC-L-alpha-phenylglycine, Boc-D-Phg-OH, N-Boc-L-2-phenylglycine

B3211-25G B3211-5G
TCB3211-25GEA 146.94 CAD
TCB3211-25G TCB3211-5G
N-(tert-Butoxycarbonyl)-L-2-phenylglycine 98,0
N-(tert-Butoxycarbonyl)-L-2-phenylglycine

CAS Number: 2900-27-8
MDL Number: MFCD00065588
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 93
Specific rotation [a]20/D: 142 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-2-phenylglycine, Boc-Phg-OH

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00065588
CAS Number: 2900-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7c40c9642e1e5d1c1a586e0a955c39bb08e68e3&vid=a7c40c9642e1e5d1c1a586e0a955c39bb08e68e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR