4-Benzyl-L-aspartate 98,0

Supplier: TCI America

Synonyms: L-Aspartic acid beta-benzyl ester, β-Benzyl L-aspartate, H-Asp(obzl)-OH, beta-Benzyl L-aspartate, L-Aspartic acid 4-benzyl ester, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, 4-Benzyl L-Aspartate, H-Asp(OBn)-OH, L-Aspartic Acid beta-Benzyl Ester, L-Aspartic acid β-benzyl ester

B3209-5G B3209-25G
TCB3209-5GEA 91.64 CAD
TCB3209-5G TCB3209-25G
4-Benzyl-L-aspartate 98,0
4-Benzyl-L-aspartate

CAS Number: 2177-63-1
MDL Number: MFCD00037208
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 221
Specific rotation [a]20/D: 27 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free), Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Aspartic Acid 4-Benzyl Ester, H-Asp(OBzl)-OH, beta-Benzyl L-Aspartate, L-Aspartic Acid beta-Benzyl Ester

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 221 °C
MDL Number: MFCD00037208
CAS Number: 2177-63-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04558eb651ddebda32c0e911e5dd319a5c59ae85&vid=04558eb651ddebda32c0e911e5dd319a5c59ae85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR