1-(2-Bromoethoxy)-2-ethoxybenzene ≥96.0%

Supplier: TCI America

B3191-25G B3191-5G
TCB3191-25GEA 211.72 CAD
TCB3191-25G TCB3191-5G
1-(2-Bromoethoxy)-2-ethoxybenzene ≥96.0%
1-(2-Bromoethoxy)-2-ethoxybenzene

CAS Number: 3259-03-8
MDL Number: MFCD02030483
Molecular Formula: C10H13BrO2
Molecular Weight: 245.12
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 43


Other Product Names / Synonyms: 2-(2-Ethoxyphenoxy)ethyl Bromide

Formula: C₁₀H₁₃BrO₂
Melting Pt: 43 °C
MDL Number: MFCD02030483
CAS Number: 3259-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bee7aed31fb55bcec6cb420d6046860eddb4a2c&vid=1bee7aed31fb55bcec6cb420d6046860eddb4a2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR