1-Benzylpiperidine-4-carbaldehyde ≥96.0% (by GC, titration analysis)
Supplier: TCI America
B3190-5G
B3190-25G
TCB3190-5GEA
104.28
CAD
TCB3190-5G
TCB3190-25G
1-Benzylpiperidine-4-carbaldehyde ≥96.0% (by GC, titration analysis)
1-Benzylpiperidine-4-carbaldehyde
CAS Number: 22065-85-6
MDL Number: MFCD02258902
Molecular Formula: C13H17NO
Molecular Weight: 203.29
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 128
Flash Point (°C): 110
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: 1-Benzyl-4-formylpiperidine
Formula:
C₁₃H₁₇NO Boiling Pt: 128 °C Density: 1.05 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD02258902 CAS Number: 22065-85-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...