5-Bromopicolinic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B3188-1G B3188-5G
TCB3188-1GEA 55.3 CAD
TCB3188-1G TCB3188-5G
5-Bromopicolinic acid ≥97.0% (by GC, titration analysis)
5-Bromopicolinic acid

CAS Number: 30766-11-1
MDL Number: MFCD00234149
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 176


Other Product Names / Synonyms: 5-Bromopicolinic Acid

Formula: C₆H₄BrNO₂
Melting Pt: 176 °C
MDL Number: MFCD00234149
CAS Number: 30766-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38aad293048cc8bef10d103c62f43fcdcffa7ba7&vid=38aad293048cc8bef10d103c62f43fcdcffa7ba7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR