(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97,0

Supplier: TCI America

Synonyms: Boc-Cis-4-F-Pro-OH, N-Boc-cis-4-fluoro-L-proline, N-(tert-Butoxycarbonyl)-cis-4-fluoro-L-proline, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid, (2S,4S)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4S)-1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acid

B3178-1G B3178-200MG
TCB3178-1GEA 239.69 CAD
TCB3178-1G TCB3178-200MG
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97,0
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

CAS Number: 203866-13-1
MDL Number: MFCD04973957
Molecular Formula: C10H16FNO4
Molecular Weight: 233.24
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 162
Specific rotation [a]20/D: -57 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Chiral Building Blocks, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: (2S,4S)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-cis-4-fluoro-L-proline, N-Boc-cis-4-fluoro-L-proline

Formula: C₁₀H₁₆FNO₄
MW: 233.24 g/mol
Melting Pt: 162 °C
MDL Number: MFCD04973957
CAS Number: 203866-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=915aa35b16cb1aadde8f648ff8942e1384742e7e&vid=915aa35b16cb1aadde8f648ff8942e1384742e7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR