(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid ≥96.0%

Supplier: TCI America

Synonyms: N-(tert-Butoxycarbonyl)-trans-4-fluoro-L-proline, (2S,4R)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, N-Boc-trans-4-fluoro-L-proline

B3177-1G B3177-200MG
TCB3177-1GEA 319 CAD
TCB3177-1G TCB3177-200MG
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid ≥96.0%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

CAS Number: 203866-14-2
MDL Number: MFCD06796085
Molecular Formula: C10H16FNO4
Molecular Weight: 233.24
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 116
Specific rotation [a]20/D: -69 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Chiral Building Blocks, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: (2S,4R)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-trans-4-fluoro-L-proline, N-Boc-trans-4-fluoro-L-proline

Formula: C₁₀H₁₆FNO₄
MW: 233.24 g/mol
Melting Pt: 116 °C
MDL Number: MFCD06796085
CAS Number: 203866-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18708576dd75ebaf6d68b70f2e979ed658cda0a0&vid=18708576dd75ebaf6d68b70f2e979ed658cda0a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR