2-Bromoindene ≥98.0%

Supplier: TCI America

B3163-5G B3163-25G
TCB3163-5GEA 105.86 CAD
TCB3163-5G TCB3163-25G
2-Bromoindene ≥98.0%
2-Bromoindene

CAS Number: 10485-09-3
MDL Number: MFCD06797863
Molecular Formula: C9H7Br
Molecular Weight: 195.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 38
Storage Temperature: 0-10°C

Formula: C₉H₇Br
Boiling Pt: 127 °C
Melting Pt: 38 °C
MDL Number: MFCD06797863
CAS Number: 10485-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9acae39befee6f0d7a02c20f386f477cecd41ec&vid=f9acae39befee6f0d7a02c20f386f477cecd41ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR