Bis(tri-tert-butylphosphine)palladium(0) ≥98.0%
Supplier: TCI America
Synonyms:
Bis(tri-tert-butylphosphine)palladium, Bis(tri-t-butylphosphine)palladium(0)
B3161-250MG
B3161-1G
TCB3161-250MGEA
136.8
CAD
TCB3161-250MG
TCB3161-1G
Bis(tri-tert-butylphosphine)palladium(0) ≥98.0%
Bis(tri-tert-butylphosphine)palladium(0)
CAS Number: 53199-31-8
MDL Number: MFCD03094580
Molecular Formula: C24H54P2Pd
Molecular Weight: 511.06
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: <0°C
Formula:
C₂₄H₅₄P₂Pd MW: 511.05 g/mol |
MDL Number:
MFCD03094580 CAS Number: 53199-31-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...