[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B3160-1G B3160-5G
TCB3160-1GEA 252.96 CAD
TCB3160-1G TCB3160-5G
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride ≥98.0% (by titrimetric analysis)
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride

CAS Number: 95408-45-0
Molecular Formula: C26H44Cl2FeP2Pd
Molecular Weight: 651.75
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), [1,1'-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II)

Formula: C₂₆H₄₄Cl₂FeP₂Pd
MDL Number: MFCD08064219
CAS Number: 95408-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73ab97fc15bb9a65679d939f7a5a63ba82d98fe5&vid=73ab97fc15bb9a65679d939f7a5a63ba82d98fe5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR