4-Bromophenyl methyl sulfone ≥98.0%
Supplier: TCI America
Synonyms:
1-Bromo-4-(methylsulphonyl)benzene, 1-Bromo-4-(methylsulfonyl)benzene
B3148-25G
B3148-5G
TCB3148-25GEA
300.2
CAD
TCB3148-25G
TCB3148-5G
4-Bromophenyl methyl sulfone ≥98.0%
4-Bromophenyl methyl sulfone
CAS Number: 3466-32-8
MDL Number: MFCD00025065
Molecular Formula: C7H7BrO2S
Molecular Weight: 235.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 104
Formula:
C₇H₇BrO₂S MW: 235.1 g/mol Melting Pt: 104 °C |
MDL Number:
MFCD00025065 CAS Number: 3466-32-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...