1-Butyl-4-chlorobenzene ≥97.0%

Supplier: TCI America

B3143-25G B3143-5G
TCB3143-25GEA 255.96 CAD
TCB3143-25G TCB3143-5G
1-Butyl-4-chlorobenzene ≥97.0%
1-Butyl-4-chlorobenzene

CAS Number: 15499-27-1
MDL Number: MFCD00051926
Molecular Formula: C10H13Cl
Molecular Weight: 168.66
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Specific Gravity (20/20): 1.00

Formula: C₁₀H₁₃Cl
MW: 168.67 g/mol
Boiling Pt: 228 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00051926
CAS Number: 15499-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1279e53d760666b567c45672a459730720cf581e&vid=1279e53d760666b567c45672a459730720cf581e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR