tert-Butyl carbamate ≥98.0%
Supplier: TCI America
B3132-25G
B3132-5G
TCB3132-25GEA
118.5
CAD
TCB3132-25G
TCB3132-5G
tert-Butyl carbamate ≥98.0%
tert-Butyl carbamate
CAS Number: 4248-19-5
MDL Number: MFCD00007962
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 109
Other Product Names / Synonyms: Carbamic Acid tert-Butyl Ester
Formula:
C₅H₁₁NO₂ Melting Pt: 109 °C |
MDL Number:
MFCD00007962 CAS Number: 4248-19-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...