4-Bromo-m-toluic acid ≥97.0%

Supplier: TCI America

Synonyms: 4-Bromo-3-methylbenzoic acid, 2-Bromo-5-carboxytoluene, 4-Bromo-m-toluic acid

B3113-25G B3113-5G
TCB3113-25GEA 480.95 CAD
TCB3113-25G TCB3113-5G
4-Bromo-m-toluic acid ≥97.0%
4-Bromo-m-toluic acid

CAS Number: 7697-28-1
MDL Number: MFCD00039526
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 215


Other Product Names / Synonyms: 4-Bromo-m-toluic Acid

Formula: C₈H₇BrO₂
Melting Pt: 215 °C
MDL Number: MFCD00039526
CAS Number: 7697-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13dc4fe724a118916a88adb1c3663ab147c471d3&vid=8c1b06207a79c86e0dc407fe0bd59d18dd0fcd64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR