2-Bromo-5-(trifluoromethoxy)phenol ≥98.0%
Supplier: TCI America
B3108-25G
B3108-5G
TCB3108-25GEA
273.34
CAD
TCB3108-25G
TCB3108-5G
2-Bromo-5-(trifluoromethoxy)phenol ≥98.0%
2-Bromo-5-(trifluoromethoxy)phenol
CAS Number: 205371-26-2
MDL Number: MFCD06660162
Molecular Formula: C7H4BrF3O2
Molecular Weight: 257.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.74
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-2-hydroxy-4-(trifluoromethoxy)benzene
Formula:
C₇H₄BrF₃O₂ Density: 1.74 (20 °C) |
MDL Number:
MFCD06660162 CAS Number: 205371-26-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...