3-Bromo-α,α,α,5-tetrafluorotoluene ≥98.0%

Supplier: TCI America

B3106-5G B3106-25G
TCB3106-5GEA 28.44 CAD
TCB3106-5G TCB3106-25G
3-Bromo-α,α,α,5-tetrafluorotoluene ≥98.0%
3-Bromo-α,α,α,5-tetrafluorotoluene

CAS Number: 130723-13-6
MDL Number: MFCD00042498
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 110
Specific Gravity (20/20): 1.71


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-3-fluoro-5-(trifluoromethyl)benzene, 3-Bromo-alpha,alpha,alpha,5-tetrafluorotoluene

Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 139 °C
Density: 1.71 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00042498
CAS Number: 130723-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6cddd9443dfc9a9a666f87773e05a7a7828cf40&vid=b6cddd9443dfc9a9a666f87773e05a7a7828cf40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR