2-Bromo-5-iodotoluene ≥98.0%
Supplier: TCI America
B3099-25G
B3099-5G
TCB3099-25GEA
363.4
CAD
TCB3099-25G
TCB3099-5G
2-Bromo-5-iodotoluene ≥98.0%
2-Bromo-5-iodotoluene
CAS Number: 202865-85-8
MDL Number: MFCD00070749
Molecular Formula: C7H6BrI
Molecular Weight: 296.93
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 2.10
Formula:
C₇H₆BrI Density: 2.10 (20 °C) |
MDL Number:
MFCD00070749 CAS Number: 202865-85-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...