2-Bromo-4-fluoro-1-iodobenzene ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-1-fluoro-4-iodobenzene, 2-Bromo-4-fluoroiodobenzene, 1-Bromo-5-fluoro-2-iodobenzene

B3098-1G B3098-5G
TCB3098-1GEA 74.26 CAD
TCB3098-1G TCB3098-5G
2-Bromo-4-fluoro-1-iodobenzene ≥98.0%
2-Bromo-4-fluoro-1-iodobenzene

CAS Number: 202865-73-4
MDL Number: MFCD00142868
Molecular Formula: C6H3BrFI
Molecular Weight: 300.90
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 110
Specific Gravity (20/20): 2.33

Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 225 °C
Density: 2.33 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00142868
CAS Number: 202865-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffdf9bced96bc4712d18c01fd69f04dd88634d39&vid=ffdf9bced96bc4712d18c01fd69f04dd88634d39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR