Benzyl phenyl sulfone ≥98.0%
Supplier: TCI America
Synonyms:
((Phenylmethyl)sulphonyl)benzol, [(Phenylmethyl)sulphonyl]benzene, Benzylsulfonylbenzene, Benzylsulphonylbenzene, (Benzylsulphonyl)benzene, (Benzylsulfonyl)benzene, ((Phenylmethyl)sulfonyl)benzol, [(Phenylmethyl)sulfonyl]benzene
B3080-25G
B3080-5G
TCB3080-25GEA
376.04
CAD
TCB3080-25G
TCB3080-5G
Benzyl phenyl sulfone ≥98.0%
Benzyl phenyl sulphone
CAS Number: 3112-88-7
MDL Number: MFCD00025040
Molecular Formula: C13H12O2S
Molecular Weight: 232.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 149
Formula:
C₁₃H₁₂O₂S Melting Pt: 149 °C |
MDL Number:
MFCD00025040 CAS Number: 3112-88-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...