N-Boc-L-prolinol ≥98.0%
Supplier: TCI America
Synonyms:
N-(tert-Butoxycarbonyl)-L-prolinol, 1-Boc-2-(S)-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol, N-Boc-L-Prolinol
CAS Number: 69610-40-8
MDL Number: MFCD00066232
Molecular Formula: C10H19NO3
Molecular Weight: 201.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 62
Specific rotation [a]20/D: -55 deg (C=5, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-L-prolinol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol Boiling Pt: 150 °C Melting Pt: 62 °C |
MDL Number:
MFCD00066232 CAS Number: 69610-40-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...