N-Boc-D-prolinol ≥98.0%

Supplier: TCI America

Synonyms: (R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, N-tert-Butoxycarbonyl-D-prolinol, N-t-Boc-D-prolinol, Boc-D-Prolinol

B3076-5G B3076-1G
TCB3076-5GEA 85.13 CAD
TCB3076-5G TCB3076-1G
N-Boc-D-prolinol ≥98.0%
N-Boc-D-prolinol

CAS Number: 83435-58-9
MDL Number: MFCD00040580
Molecular Formula: C10H19NO3
Molecular Weight: 201.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62
Specific rotation [a]20/D: 54 deg (C=5, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-D-prolinol, (R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-1-Boc-2-pyrrolidinemethanol

Formula: C₁₀H₁₉NO₃
MW: 201.3 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00040580
CAS Number: 83435-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b60dffa78bfc2c0c8815fa10df021381c4aaf74&vid=1b60dffa78bfc2c0c8815fa10df021381c4aaf74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR